missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl adipate, 99%

CAS: 627-93-0 | C8H14O4 | 174.196 g/mol

Supplier:  Thermo Scientific Chemicals B2117418

 View more versions of this product

Catalog No. AAB2117418


Only null left
Explore available promotions

Description

Description

Dimethyl adipate is used in cosmetics (in emollients and skin conditioning). It acts as a cosmetic plasticizer. It is also used in agrochemicals and dye as well as a precursor for the preparation of active pharmaceutical ingredients. Further, it serves as a polymer intermediate. In addition to this, it is employed as a solvent for paint stripping and resins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethyl adipate is used in cosmetics (in emollients and skin conditioning). It acts as a cosmetic plasticizer. It is also used in agrochemicals and dye as well as a precursor for the preparation of active pharmaceutical ingredients. Further, it serves as a polymer intermediate. In addition to this, it is employed as a solvent for paint stripping and resins.

Solubility
Miscible with alcohols and ether. Immiscible with water.

Notes
Moisture sensitive. Incompatible with acids, bases, oxidizing agents and reducing agents.
Specifications

Specifications

Dimethyl adipate
8°C
109°C to 110°C (14 mmHg)
C8H14O4
MFCD00008469
1707443
dimethyl adipate, dimethyladipate, adipic acid dimethyl ester, hexanedioic acid, dimethyl ester, adipic acid, dimethyl ester, 1,6-dimethylhexanedioate, methyl adipate van, 1,6-dimethyl hexanedioate, dbe 6, unii-by71rx0r62
UDSFAEKRVUSQDD-UHFFFAOYSA-N
dimethyl hexanedioate
12329
174.2
627-93-0
1.063
107°C (224°F)
1.428
50 g
14,162
Miscible with alcohols and ether. Immiscible with water.
COC(=O)CCCCC(=O)OC
174.196
CHEBI:34715
99%

Chemical Identifiers

627-93-0
174.196
UDSFAEKRVUSQDD-UHFFFAOYSA-N
12329
dimethyl hexanedioate
C8H14O4
MFCD00008469
dimethyl adipate, dimethyladipate, adipic acid dimethyl ester, hexanedioic acid, dimethyl ester, adipic acid, dimethyl ester, 1,6-dimethylhexanedioate, methyl adipate van, 1,6-dimethyl hexanedioate, dbe 6, unii-by71rx0r62
CHEBI:34715
COC(=O)CCCCC(=O)OC
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 211-020-6

RTECSNumber : AV1645000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only