Learn More
Dimethyl adipate, 99%
Description
Dimethyl adipate is used in cosmetics (in emollients and skin conditioning). It acts as a cosmetic plasticizer. It is also used in agrochemicals and dye as well as a precursor for the preparation of active pharmaceutical ingredients. Further, it serves as a polymer intermediate. In addition to this, it is employed as a solvent for paint stripping and resins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Dimethyl adipate is used in cosmetics (in emollients and skin conditioning). It acts as a cosmetic plasticizer. It is also used in agrochemicals and dye as well as a precursor for the preparation of active pharmaceutical ingredients. Further, it serves as a polymer intermediate. In addition to this, it is employed as a solvent for paint stripping and resins.
Solubility
Miscible with alcohols and ether. Immiscible with water.
Notes
Moisture sensitive. Incompatible with acids, bases, oxidizing agents and reducing agents.
Specifications
Specifications
| Melting Point | 8°C |
| Density | 1.063 |
| Boiling Point | 109°C to 110°C (14 mmHg) |
| Flash Point | 107°C (224°F) |
| Refractive Index | 1.428 |
| Quantity | 50 g |
| Beilstein | 1707443 |
| Merck Index | 14,162 |
| Solubility Information | Miscible with alcohols and ether. Immiscible with water. |
| Formula Weight | 174.2 |
| Show More |
Safety and Handling
| EINECSNumber | 211-020-6 |
| RTECSNumber | AV1645000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only