accessibility menu, dialog, popup

UserName

CAS RN 627-93-0

IUPAC Name: 1,6-dimethyl hexanedioate
Synonyms: dimethyl adipate 1,6-dimethylhexanedioate dbe 6
Molecular Weight (g/mol): 174.2
Molecular Formula: C8H14O4
InChi Key: UDSFAEKRVUSQDD-UHFFFAOYSA-N
SMILES: COC(=O)CCCCC(=O)OC
Boiling Point 
  • (3)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (2)
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (5)
  • (3)
  • (2)
  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)