missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl 5-Bromoisophthalate 98.0+%, TCI America™

Catalog No. D40681G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D40681G 1 g
D40685G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D40681G Supplier TCI America Supplier No. D40681G

Chemical Identifiers

CAS 51760-21-5
Molecular Formula C10H9BrO4
Molecular Weight (g/mol) 273.082
MDL Number MFCD00078709
InChI Key QUJINGKSNJNXEB-UHFFFAOYSA-N
PubChem CID 103954
IUPAC Name dimethyl 5-bromobenzene-1,3-dicarboxylate
SMILES COC(=O)C1=CC(=CC(=C1)Br)C(=O)OC

Specifications

Melting Point 89°C
Color White-Yellow
Boiling Point 159°C
Quantity 1 g
Synonym dimethyl 5-bromoisophthalate, 5-bromoisophthalic acid dimethyl ester, dimethyl-5-bromoisophthalate, 1,3-benzenedicarboxylic acid, 5-bromo-, dimethyl ester, 1,3-dimethyl 5-bromobenzene-1,3-dicarboxylate, dimethyl5-bromoisophthalate, 5-bromo-isophthalic acid dimethyl ester, 1,3-benzenedicarboxylic acid, 5-bromo-, 1,3-dimethyl ester, methyl 5-bromo-3-methoxycarbonyl benzoate, 5-bromoisophthalic acid, dimethyl ester
Formula Weight 273.08
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Dimethyl 5-Bromoisophthalate