missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl 2-hydroxyethylphosphonate, 95%, Thermo Scientific™

Identifiants chimiques

CAS 54731-72-5
Molecular Formula C4H11O4P
Molecular Weight (g/mol) 154.10
MDL Number MFCD00043376
InChI Key TZPPDWDHNIMTDQ-UHFFFAOYSA-N
Synonym dimethyl 2-hydroxyethylphosphonate, dimethyl 2-hydroxyethyl phosphonate, 2-hydroxyethylphosphonic acid dimethyl ester, 2-hydroxyethyl phosphonic acid dimethyl ester, acmc-1ao1t, 2-diethylphosphono ethan-1-ol, tzppdwdhnimtdq-uhfffaoysa, hydroxyethyldimethylphosphonic acid, 2-dimethoxyphosphoryl ethan-1-ol
PubChem CID 2733833
IUPAC Name 2-dimethoxyphosphorylethanol
SMILES COP(=O)(CCO)OC
voir les spécifications

Produits 2
Numéro de catalogue Numéro du manufacturier. Quantity Packaging Prix Quantity & Availability  
Numéro de catalogue Numéro du manufacturier. Quantity Packaging Prix Quantity & Availability  
AC338160050
Afficher les documents Détails de la promotion
N/A 5mL Glass bottle N/A
N/A
Pour de laide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
AC338160250
Afficher les documents Détails de la promotion
N/A 25mL Glass bottle N/A
N/A
Pour de laide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

for C-C Bond Formation (Olefination)
Spécifications

Identifiants chimiques

54731-72-5
154.10
TZPPDWDHNIMTDQ-UHFFFAOYSA-N
2733833
COP(=O)(CCO)OC
C4H11O4P
MFCD00043376
dimethyl 2-hydroxyethylphosphonate, dimethyl 2-hydroxyethyl phosphonate, 2-hydroxyethylphosphonic acid dimethyl ester, 2-hydroxyethyl phosphonic acid dimethyl ester, acmc-1ao1t, 2-diethylphosphono ethan-1-ol, tzppdwdhnimtdq-uhfffaoysa, hydroxyethyldimethylphosphonic acid, 2-dimethoxyphosphoryl ethan-1-ol
2-dimethoxyphosphorylethanol

Spécifications

54731-72-5
98.0°C to 104.0°C (0.1mmHg)
Authentic
C4H11O4P
(CH3O)2P(O)(CH2)2OH
04, IV, 3584
dimethyl 2-hydroxyethylphosphonate, dimethyl 2-hydroxyethyl phosphonate, 2-hydroxyethylphosphonic acid dimethyl ester, 2-hydroxyethyl phosphonic acid dimethyl ester, acmc-1ao1t, 2-diethylphosphono ethan-1-ol, tzppdwdhnimtdq-uhfffaoysa, hydroxyethyldimethylphosphonic acid, 2-dimethoxyphosphoryl ethan-1-ol
TZPPDWDHNIMTDQ-UHFFFAOYSA-N
2-dimethoxyphosphorylethanol
2733833
95%
1.2470g/mL
192°C
94% min. (GC)
1.4430 to 1.4460
MFCD00043376
1.247
Solubility in water: soluble. Other solubilities: soluble in most common organic solvents
COP(=O)(CCO)OC
154.10
154.1
Dimethyl 2-hydroxyethylphosphonate, 95%
Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 259-310-1

FDS
Documents

Documents

RUO – Research Use Only