missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl 1,4-Cyclohexanedione-2,5-dicarboxylate 98.0+%, TCI America™

Catalog No. C1343500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C1343500G 500 g
C134325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C1343500G Supplier TCI America Supplier No. C1343500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6289-46-9
Molecular Formula C10H12O6
Molecular Weight (g/mol) 228.20
MDL Number MFCD00001607
InChI Key MHKKFFHWMKEBDW-UHFFFAOYNA-N
Synonym dimethyl succinylsuccinate, dimethyl 1,4-cyclohexanedione-2,5-dicarboxylate, succinosuccinic acid, dimethyl ester, dmss, dimethyl succinylosuccinate, dimethyl 2,5-dioxo-1,4-cyclohexanedicarboxylate, dimethyl succinylo succinate, 1,4-cyclohexanedicarboxylic acid, 2,5-dioxo-, dimethyl ester, 2,5-dioxo-1,4-cyclohexanedicarboxylic acid dimethyl ester, 1,4-cyclohexanedicarboxylic acid, 2,5-dioxo-, 1,4-dimethyl ester
PubChem CID 94866
IUPAC Name 1,4-dimethyl 2,5-dioxocyclohexane-1,4-dicarboxylate
SMILES COC(=O)C1CC(=O)C(CC1=O)C(=O)OC

Specifications

Melting Point 156°C
Quantity 500 g
Formula Weight 228.20
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Dimethyl 1,4-Cyclohexanedione-2,5-dicarboxylate