missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl 1,4-Cyclohexanedicarboxylate (cis- and trans- mixture) 98.0+%, TCI America™

Catalog No. C1154500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C1154500G 500 g
C115425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C1154500G Supplier TCI America Supplier No. C1154500G

Chemical Identifiers

CAS 94-60-0
Molecular Formula C10H16O4
Molecular Weight (g/mol) 200.234
MDL Number MFCD00001460
InChI Key LNGAGQAGYITKCW-UHFFFAOYSA-N
Synonym dimethyl 1,4-cyclohexanedicarboxylate, dimethyl trans-1,4-cyclohexanedicarboxylate, 1r,4r-dimethyl cyclohexane-1,4-dicarboxylate, cis-dimethyl cyclohexane-1,4-dicarboxylate, dimethyl trans-cyclohexane-1,4-dicarboxylate, 1,4-cyclohexanedicarboxylic acid, dimethyl ester, dimethyl hexahydroterephthalate, trans-dimethyl cyclohexane-1,4-dicarboxylate, 1,4-cyclohexanedicarboxylic acid, dimethyl ester, trans, unii-ys87mds0zv
PubChem CID 7198
IUPAC Name dimethyl cyclohexane-1,4-dicarboxylate
SMILES COC(=O)C1CCC(CC1)C(=O)OC

Specifications

Color Colorless
Boiling Point 263°C
Quantity 500 g
Formula Weight 200.23
Percent Purity ≥92.0% (GC)
Physical Form Liquid
Chemical Name or Material Dimethyl 1,4-Cyclohexanedicarboxylate (cis- and trans- mixture)
EINECSNumber (3)-2449
TSCA Yes