missing translation for 'onlineSavingsMsg'
Learn More

Interscience DiluFlow Nozzles

Set of 5 nozzles for use with DiluFlow.

Supplier:  Interscience 513040

 Afficher plus de versions de ce produit

Numéro de catalogue. 16949020


Il en reste null

Description

Description

  • Set of 5 nozzles and caps for DiluFlow tubings

PTFE set:

  • Fits tubings with inner diameter of 4.8 to 8 mm

PEEK set:

  • Fits tubings with inner diameter of 1.6 to 3.2 mm
Spécifications

Identifiants chimiques

Tubing Nozzle
Not Autoclavable
Non-sterile
5/Pk.

Spécifications

Tubing Nozzle
1.6 to 3.2 mm dia. Tubings
5/Pk.
Not Autoclavable
Non-sterile
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions