missing translation for 'onlineSavingsMsg'
Learn More

Interscience DiluFlow Dispensing Assembly

Numéro de catalogue. 16948935
Change view
Click to view available options
For Use With (Equipment):
3.2 mm Tubings
6.4 mm Tubings
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. For Use With (Equipment)
16948935 6.4 mm Tubings
16948936 3.2 mm Tubings
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. 16948935 Fournisseur Interscience Code fournisseur 513043
Il en reste null

Dispensing assembly for DiluFlow.

Includes:

  • Set of silicone tubings
  • GL45 screw cap
  • Tube weight
  • Nozzle
  • Filter

Available for 3.2 mm or 6.4 mm tubings

Spécifications

Product Type Dispensing Tubing
Autoclavable Autoclavable
For Use With (Equipment) 6.4 mm Tubings
Sterility Non-sterile