missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dilithium tetrachlorocuprate, 0.1M solution in THF
CAS: 15489-27-7 | Cl4CuLi2 | 219.226 g/mol
$167.15 - $167.15
Specifications
| Chemical Name or Material | Dilithium tetrachlorocuprate |
|---|---|
| Color | Colorless |
| Name Note | 0.1M Solution in THF |
| CAS | 15489-27-7 , 109-99-9 |
| Molecular Formula | Cl4CuLi2 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC407831000
|
Thermo Scientific Chemicals
407831000 |
100 mL |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Dilithium tetrachlorocuprate | |
| 0.1M Solution in THF | |
| Cl4CuLi2 | |
| MFCD00011081 | |
| 04,163; 05,226; 06,203; 07,114; 08,176; 11,190; 12,195 | |
| HCJWWBBBSCXJMS-UHFFFAOYSA-J | |
| dilithium;tetrachlorocopper(2-) | |
| 11074879 | |
| Glass bottle | |
| −17°C |
| Colorless | |
| 15489-27-7 , 109-99-9 | |
| Li2CuCl4 | |
| 0.09 to 0.12M | |
| 0.9100g/mL | |
| [Li+].[Li+].Cl[Cu-2](Cl)(Cl)Cl | |
| 219.226 | |
| 219.24 | |
| 0.91 | |
| Liquid |
RUO – Research Use Only