CAS RN 15489-27-7
CAS RN 15489-27-7
IUPAC Name:
copper(2+) dilithium(1+) tetrachloride
Synonyms:
copper(2+) dilithium(1+) tetrachloride
Molecular Weight (g/mol):
219.23
Molecular Formula:
Cl4CuLi2
InChi Key:
HCJWWBBBSCXJMS-UHFFFAOYSA-J
SMILES:
[Li+].[Li+].[Cl-].[Cl-].[Cl-].[Cl-].[Cu++]
1
–
4
of
4
results
2
Dilithium tetrachlorocuprate, 0.1M solution in THF
CAS: 15489-27-7 | Cl4CuLi2 | 219.226 g/mol
3
Lithium tetrachlorocuprate, 0.1M in THF
CAS: 15489-27-7 Molecular Formula: Cl4CuLi2 Molecular Weight (g/mol): 219.226 MDL Number: MFCD00011081 InChI Key: HCJWWBBBSCXJMS-UHFFFAOYSA-J PubChem CID: 11074879 IUPAC Name: dilithium;tetrachlorocopper(2-) SMILES: [Li+].[Li+].Cl[Cu-2](Cl)(Cl)Cl
4
Dilithium Tetrachlorocopper(II) (ca. 2.5% in Tetrahydrofuran, ca. 0.1mol/L), TCI America™
CAS: 15489-27-7 Molecular Formula: Cl4CuLi2 Molecular Weight (g/mol): 219.226 MDL Number: MFCD00011081 InChI Key: HCJWWBBBSCXJMS-UHFFFAOYSA-J Synonym: Dilithium Tetrachlorocuprate(II) PubChem CID: 11074879 IUPAC Name: dilithium;tetrachlorocopper(2-) SMILES: [Li+].[Li+].Cl[Cu-2](Cl)(Cl)Cl