missing translation for 'onlineSavingsMsg'
Learn More

Diisopropylamine Dichloroacetate 98.0+%, TCI America™

Catalog No. D50745G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D50745G 5 g
D50741G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D50745G Supplier TCI America Supplier No. D50745G

Chemical Identifiers

CAS 660-27-5
Molecular Formula C8H17Cl2NO2
Molecular Weight (g/mol) 230.13
MDL Number MFCD00868288
InChI Key ILKBHIBYKSHTKQ-UHFFFAOYSA-N
Synonym diisopropylamine dichloroacetate, diisopropylammonium dichloroacetate, diisopropylamine 2,2-dichloroacetate, vasculopatina, dipromonium, cubisol, dapocel, disotat, kalodil, oxypangam
PubChem CID 12617
IUPAC Name 2,2-dichloroacetic acid; bis(propan-2-yl)amine
SMILES OC(=O)C(Cl)Cl.CC(C)NC(C)C

Specifications

Melting Point 122°C
Color White
Quantity 5 g
Formula Weight 230.13
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Diisopropylamine Dichloroacetate
RTECSNumber AG6475000
TSCA Yes