missing translation for 'onlineSavingsMsg'
Learn More

Diisopropylamine, 99+%, Thermo Scientific Chemicals

Catalog No. AC219432500
Change view
Click to view available options
Quantity:
250 mL
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC219432500 250 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC219432500 Supplier Thermo Scientific Chemicals Supplier No. 219432500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 108-18-9
Molecular Weight (g/mol) 101.19
MDL Number MFCD00008862
InChI Key UAOMVDZJSHZZME-UHFFFAOYSA-N
Synonym diisopropylamine, 2-propanamine, n-1-methylethyl, n-1-methylethyl-2-propanamine, dipa, n,n-diisopropylamine, bis propan-2-yl amine, n-isopropylpropan-2-amine, di-isopropylamine, n-isopropyl-1-amino-2-methylethane, diisopropyl amine
PubChem CID 7912
SMILES CC(C)NC(C)C

Specifications

Melting Point -61.0°C
Density 0.7160g/mL
Boiling Point 84.0°C
Flash Point −17°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Molecular Formula C6H15N
Refractive Index 1.3910 to 1.3930
Linear Formula [(CH3)2CH]2NH
Quantity 250 mL
Beilstein 04, 154
Specific Gravity 0.716
Merck Index 15, 3215
Solubility Information Solubility in water: 100g/L (20°C). Other solubilities: soluble in alcohol,soluble in most common organic solvents
IUPAC Name N-propan-2-ylpropan-2-amine
Viscosity 0.4 mPa.s (20°C)
Formula Weight 101.19
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Diisopropylamine, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Toxic if inhaled.
May cause respiratory irritation.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
EINECSNumber 203-558-5
RTECSNumber IM4025000
TSCA TSCA

RUO – Research Use Only