missing translation for 'onlineSavingsMsg'
Learn More

Diisopropylamine, 99%, Spectrum™ Chemical

Catalog No. 18610208
Change view
Click to view available options
Quantity:
4 L
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18610208 4 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18610208 Supplier Spectrum Chemical Mfg Cor Supplier No. D22824LTGL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 108-18-9
Molecular Formula C6H15N
Molecular Weight (g/mol) 101.19
InChI Key UAOMVDZJSHZZME-UHFFFAOYSA-N
IUPAC Name bis(propan-2-yl)amine
SMILES CC(C)NC(C)C

Specifications

CAS Max % 1
Boiling Point 84°C / 183.2°F
Packaging Amber Glass Bottle
Quantity 4 L
Percent Purity 0.99
Grade Reagent
Physical Form Liquid
Chemical Name or Material Diisopropylamine

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.