missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diisopropyl phthalate, Certified Reference Material, MilliporeSigma™ Supelco™
Spécifications
Spécifications
| CAS | 605-45-8 |
| Product Line | TraceCERT |
| Density | 1.063 g/mL (at 20°C) |
| Linear Formula | C14H18O4 |
| MDL Number | MFCD00053717 |
| Synonym | 1,2-Benzenedicarboxylic acid 1,2-bis(1-methylethyl) ester; NSC 15315; Phthalic acid diisopropyl ester |
| Quantity | 50 mg |
| Formula Weight | 250.29 |
| Grade | TraceCERT™ Analytical Standard |
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC