missing translation for 'onlineSavingsMsg'
Learn More

(+)-Diisopropyl L-tartrate, 97%

Catalog No. AAA1694122
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1694122 100 g
AAA1694114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1694122 Supplier Thermo Scientific Chemicals Supplier No. A1694122
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2217-15-4 | C10H18O6 | 234.248 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2217-15-4
Molecular Formula C10H18O6
Molecular Weight (g/mol) 234.248
MDL Number MFCD00064450
InChI Key XEBCWEDRGPSHQH-HTQZYQBOSA-N
Synonym +-diisopropyl l-tartrate, 2r,3r-diisopropyl 2,3-dihydroxysuccinate, di-isopropyl tartrate, diisoprropyl-l-tartrate, +-dipt, l-+-tartaric acid diisopropyl ester, diisopropyl l-tartrate, l-+-diisopropyl-tartrate, l-dipt, butanedioic acid, 2,3-dihydroxy-, bis 1-methylethyl ester, r,r-+/-, 1,4-diisopropyl 2r,3r-2,3-dihydroxybutanedioate
PubChem CID 6453580
IUPAC Name dipropan-2-yl (2R,3R)-2,3-dihydroxybutanedioate
SMILES CC(C)OC(=O)C(C(C(=O)OC(C)C)O)O

Specifications

Density 1.117
Boiling Point 150°C to 151°C (11 mmHg)
Flash Point 110°C (230°F)
Refractive Index 1.44
Quantity 100 g
Beilstein 1727863
Optical Rotation +16° (Neat)
Formula Weight 234.25
Percent Purity ≥98%
Chemical Name or Material (+)-Diisopropyl L-tartrate
EINECSNumber 218-709-0
RTECSNumber WW8100000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only