missing translation for 'onlineSavingsMsg'
Learn More

Diisopropyl Ether, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111023043
Change view
Click to view available options
Quantity:
20 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111023043 20 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111023043 Supplier Merck Emd Millipore Supplier No. RTC00008520ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control. Provides pharma laboratories and manufacturers with a convenient, and cost-effective alternative to the preparation of in-house working standards.

Specifications

CAS 108-20-3
Melting Point -85°C (literature)
Density 0.725 g/mL (at 25°C (literature))
Boiling Point 68°C to 69°C (literature)
Flash Point -28°C (Not Applicable)
Molecular Formula C6H14O
Linear Formula (CH3)2 CHOCH(CH3)2
Refractive Index n20/D 1.367 (literature)
Quantity 20 mL
MDL Number MFCD00008880
Synonym Diisopropyl ether; Isopropyl ether
Vapor Density 3.5 (vs air)
Formula Weight 102.17
Vapor Pressure 120 mmHg (20°C)
Grade Certified Reference Material
Show More Show Less