missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diisodecyl 4-Cyclohexene-1,2-dicarboxylate 97.0+%, TCI America™
Specifications
Specifications
| Color | Colorless |
| Quantity | 500 mL |
| Formula Weight | 450.70 |
| Percent Purity | ≥97.0% (T) |
| Physical Form | Liquid |
| Chemical Name or Material | Diisodecyl 4-Cyclohexene-1,2-dicarboxylate |
Safety and Handling
| EINECSNumber | (3)-2437 |
| TSCA | No |