missing translation for 'onlineSavingsMsg'
Learn More

Diisobutylamine, TCI America™

Catalog No. D061425ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D061425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D061425ML Supplier TCI America Supplier No. D061425ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 110-96-3
Molecular Formula C8H20N
Molecular Weight (g/mol) 130.25
MDL Number MFCD00008930
InChI Key OBYVIBDTOCAXSN-OCAPTIKFSA-O
Synonym diisobutylamine, 1-propanamine, 2-methyl-n-2-methylpropyl, bis 2-methylpropyl amine, amine, diisobutyl, bis beta-methylpropyl amine, di-isobutylamine, n,n-bis 2-methylpropyl amine, unii-t18y0a819s, ccris 6232, di-2-methylpropyl amine
PubChem CID 8085
IUPAC Name [(2R)-butan-2-yl][(2S)-butan-2-yl]azanium
SMILES CC[C@H](C)[NH2+][C@H](C)CC

Specifications

Melting Point -74°C
Color Colorless
Boiling Point 139°C
Quantity 25 mL
UN Number 2361
Formula Weight 129.25
Physical Form Liquid
Chemical Name or Material Diisobutylamine
EINECSNumber (2)-0137
RTECSNumber TX1750000
TSCA Yes