missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diisobutylaluminium hydride, 1.1M solution in cyclohexane, AcroSeal™
CAS: 1191-15-7 | C8H19Al
Supplier: Thermo Scientific Chemicals 201031000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid, Reduction reagentSpecifications
| Diisobutylaluminium hydride | |
| 1.1M Solution in Cyclohexane | |
| -70.0°C | |
| [(CH3)2CHCH2]2AIH | |
| 04, IV, 4400 | |
| 01,260; 02,140; 03,101; 04,158; 05,224; 06,198; 07,111; 08,173; 09,171; 10,149; 11,185; 12,191; 13,115; 14,138; 15,137; 16,27; 17,123 | |
| 15, 3212 | |
| Solubility in water: vigorous reaction. Other solubilities: miscible with saturated aliphatic and aromatic, hydrocarbons | |
| AcroSeal™ Glass bottle | |
| 100 mL | |
| Solution |
RUO – Research Use Only