accessibility menu, dialog, popup

UserName

CAS RN 1191-15-7

H3CCH3AlCH3CH3

CAS RN 1191-15-7

IUPAC Name: bis(2-methylpropyl)alumanyl
Synonyms: diisobutylaluminium hydride
Molecular Weight (g/mol): 141.21
Molecular Formula: C8H18Al
InChi Key: SIPUZPBQZHNSDW-UHFFFAOYSA-N
SMILES: CC(C)C[Al]CC(C)C
Quantity 
  • (4)
  • (2)
  • (2)
  • (2)
Physical Form 
  • (6)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results