missing translation for 'onlineSavingsMsg'
Learn More

Diisobutylaluminium hydride, 1.1M solution in cyclohexane, AcroSeal™

CAS: 1191-15-7 | C8H19Al

$135.08 - $135.08

Specifications

Chemical Name or Material Diisobutylaluminium hydride
Color Undesignated
Name Note 1.1M Solution in Cyclohexane
CAS 1191-15-7 , 110-82-7
Melting Point -70.0°C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC201031000
View Documents
Thermo Scientific Chemicals
201031000
100 mL AcroSeal™ Glass bottle
Each for $135.08
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid, Reduction reagent
Specifications

Specifications

Diisobutylaluminium hydride
1.1M Solution in Cyclohexane
-70.0°C
[(CH3)2CHCH2]2AIH
04, IV, 4400
01,260; 02,140; 03,101; 04,158; 05,224; 06,198; 07,111; 08,173; 09,171; 10,149; 11,185; 12,191; 13,115; 14,138; 15,137; 16,27; 17,123
15, 3212
Solubility in water: vigorous reaction. Other solubilities: miscible with saturated aliphatic and aromatic, hydrocarbons
0.78
Solution
Undesignated
1191-15-7 , 110-82-7
C8H19Al
MFCD00008928
19.5 to 20.5% (DIBAL-H, on Al basis)
0.7800g/mL
DIBAL-H, 1.1M solution in cyclohexane
142.22
−018°C
Videos
SDS
Documents

Documents

RUO – Research Use Only