missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diisobutylaluminium hydride, 1.0M solution in heptane, AcroSeal™
CAS: 1191-15-7 | C8H19Al
$132.32 - $132.32
Specifications
| Chemical Name or Material | Diisobutylaluminium hydride |
|---|---|
| Color | Undesignated |
| Name Note | 1.0M Solution in Heptane |
| CAS | 1191-15-7 , 142-82-5 |
| Melting Point | -70.0°C |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC201051000
|
Thermo Scientific Chemicals
201051000 |
100 mL | AcroSeal™ Glass bottle |
Each for $132.32
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid, Reduction reagentSpecifications
| Diisobutylaluminium hydride | |
| 1.0M Solution in Heptane | |
| -70.0°C | |
| [(CH3)2CHCH2]2AIH | |
| 04, IV, 4400 | |
| 01,260; 02,140; 03,101; 04,158; 05,224; 06,198; 07,111; 08,173; 09,171; 10,149; 11,185; 12,191; 13,115; 14,138; 15,137; 16,27; 17,123 | |
| 15, 3212 | |
| Solubility in water: vigorous reaction. Other solubilities: miscible with saturated aliphatic and aromatic, hydrocarbons | |
| 0.7 | |
| Solution |
RUO – Research Use Only