missing translation for 'onlineSavingsMsg'
Learn More

Diisobutyl Phosphite 95.0+%, TCI America™

Catalog No. D3417500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D3417500G 500 g
D341725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D3417500G Supplier TCI America Supplier No. D3417500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1189-24-8
Molecular Formula C8H18O3P+
Molecular Weight (g/mol) 193.203
InChI Key QPKOILOWXGLVJS-UHFFFAOYSA-N
Synonym Phosphorous Acid Diisobutyl Ester, Diisobutyl Phosphonate, Phosphonic Acid Diisobutyl Ester
PubChem CID 6327151
IUPAC Name bis(2-methylpropoxy)-oxophosphanium
SMILES CC(C)CO[P+](=O)OCC(C)C

Specifications

Color Yellow
Boiling Point 138°C
Quantity 500 g
Formula Weight 194.21
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Diisobutyl Phosphite