Learn More
Dihydrogen hexachloroplatinate(IV) hexahydrate, ACS, Premion™, 99.95% (metals basis), Pt 37.5% min
Description
Dihydrogen hexachloroplatinate(IV) hexahydrate is used for the determination of potassium. It is also used for the purification of platinum by treating it with ammonium chloride flowed by heating of ammonium hexachloroplatinate with hydrogen yields elemental platinum. It also finds application in catalysis, electroplating, photography, platinum mirrors, printing for etching of zinc, producing fine color in glass and porcelain, indelible ink and microscopy. It is a precursor for the preparation of other platinum catalysts.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 60°C (decomposition) |
| Color | Red to Brown |
| Physical Form | Lump |
| Quantity | 1 g |
| Sensitivity | Hygroscopic |
| Appearance | Red-brown lump |
| Assay Percent Range | 99.95% (metals basis) |
| Density | 2.43 |
| Solubility Information | Soluble in water,ethanol,ether,ethyl acetate and acetone. Insoluble in nitric acid. |
| Formula Weight | 517.91 (409.82 Anhydrous) |
| Show More |
Safety and Handling
| Hazard Category | H301-H314-H317-H334-H335 |
| Precautionary Statement | P260-P264b-P270-P271-P272-P280-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c |
| DOTInformation | DOT Class: 8; Packing Group: III |
| EINECSNumber | 241-010-7 |
| RTECSNumber | TP1500000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only