missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dihydrocapsaicin, ≥97.0% (HPLC), MilliporeSigma™ Supelco™
Description
Cleaning Products, Cosmetics, Food and Beverages and Personal Care
Specifications
Specifications
| CAS | 19408-84-5 |
| Linear Formula | C18H29 NO3 |
| MDL Number | MFCD00057794 |
| Synonym | 6,7-Dihydrocapsaicin; 8-Methyl-N-vanillylnonanamide; N-[(4-Hydroxy-3-methoxyphenyl)methyl]-8-methylnonenamide |
| Type | Analytical Standard |
| Quantity | 5 mg |
| Formula Weight | 307.43 |
| Percent Purity | ≥97.0% (HPLC) |
| Grade | Analytical Standard |