missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Digoxin, ≥95% (HPLC), MilliporeSigma™ Supelco™
Supplier: Merck Emd Millipore D60031G
Description
Featured Industries: Food and BeveragesSpecifications
| 20830-75-5 | |
| Neat | |
| MFCD00003674 | |
| Analytical Standard | |
| 780.94 | |
| Analytical Standard |
| Ambient | |
| C41H64O14 | |
| 12 beta-Hydroxydigitoxin | |
| 1 g | |
| ≥95% (HPLC) |