missing translation for 'onlineSavingsMsg'
Learn More

Digoxin, ≥95% (HPLC), MilliporeSigma™ Supelco™

Catalog No. p-7187399
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020499 100 mg
111018693 1 g
111018014 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 111020499 Supplier Merck Emd Millipore Supplier No. D6003100MG
Only null left
Featured Industries: Food and Beverages

Specifications

CAS 20830-75-5
Shipping Condition Ambient
Physical Form Neat
Linear Formula C41H64O14
MDL Number MFCD00003674
Synonym 12 beta-Hydroxydigitoxin
Type Analytical Standard
Quantity 100 mg
Formula Weight 780.94
Percent Purity ≥95% (HPLC)
Grade Analytical Standard
Show More Show Less