accessibility menu, dialog, popup

UserName

H3C(R)O(S)(S)HO(S)HOO(S)(S)OH(S)O(R)H3CO(S)(S)OH(R)O(R)H3CO(S)(S)H3C(R)(R)(S)(R)HO(S)H3C(R)(S)OHOO

CAS RN 20830-75-5

IUPAC Name: 4-[(1R,3aS,3bR,5aR,7S,9aS,9bS,11R,11aS)-7-{[(2R,4S,5S,6R)-5-{[(2S,4S,5S,6R)-5-{[(2S,4S,5S,6R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy}-4-hydroxy-6-methyloxan-2-yl]oxy}-4-hydroxy-6-methyloxan-2-yl]oxy}-3a,11-dihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]-2,5-dihydrofuran-2-one
Synonyms: digoxin 地高辛 digitalis glycoside 12β-hydroxydigitoxin
Molecular Weight (g/mol): 780.95
Molecular Formula: C41H64O14
InChi Key: LTMHDMANZUZIPE-PUGKRICDSA-N
SMILES: C[C@H]1O[C@H](C[C@H](O)[C@@H]1O)O[C@H]1[C@@H](O)C[C@H](O[C@H]2[C@@H](O)C[C@H](O[C@H]3CC[C@@]4(C)[C@H](CC[C@@H]5[C@@H]4C[C@@H](O)[C@]4(C)[C@H](CC[C@]54O)C4=CC(=O)OC4)C3)O[C@@H]2C)O[C@@H]1C
Molecular Weight (g/mol) 
  • (4)
  • (4)
Percent Purity 
  • (3)
  • (2)
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
Grade 
  • (3)
  • (1)
Industry Type 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (4)
  • (2)
  • (3)
  • (4)
  • (4)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)