Learn More
United Scientific™ Digital Sound Level Meter
Monitor sound levels effortlessly with the Digital Sound Level Meter, ideal for studying noise pollution in various environments.
Supplier: United Scientific™ DSNDLM
Avertissement:
Cancer and reproductive harm.
Description
- Auto power-off
- Measures 30 to 130 dBA
- Backlight display
Spécifications
| Digital Sound Level Meter |