missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Difluoroacetic Acid 97.0+%, TCI America™
Specifications
Specifications
| Melting Point | 0°C |
| Boiling Point | 135°C |
| Quantity | 25 g |
| UN Number | 3265 |
| Formula Weight | 96.03 |
| Percent Purity | ≥97.0% (T) |
| Physical Form | Liquid |
| Chemical Name or Material | Difluoroacetic Acid |
Safety and Handling
| RTECSNumber | AG9900000 |
| TSCA | Yes |