missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Difco™ Yeast Extract, low-dusting (LD)

Formulation of culture media for cultivation of heterotrophic microorganisms
$285.61 - $4,271.25
Specifications
| Content And Storage | Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers. |
|---|---|
| Cell Type | Bacteria |
| Culture Type | Mammalian, Insect, Microbial |
| Form | Powder |
| Product Type | Dehydrated Culture Media |
Description
- Yeast extract designed to limit the amount of dust generated compared to other spray-dried yeast extracts
- Yeast Extracts are concentrates of the water-soluble portion of autolyzed Saccharomyces cerevisiae cells
- Gibco™ yeast extracts are derived from primary grown baker’s yeast
- These animal origin-free products are suitable for use as multi-functional nutritional supplements in mammalian cell culture, microbial fermentation, and insect cell culture applications
- Yeast extract provides essential water-soluble vitamins, amino acids, peptides, and carbohydrates to any medium formulation
Specifications
| Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers. | |
| Mammalian, Insect, Microbial | |
| Dehydrated Culture Media | |
| Yeast, animal origin-free |
| Bacteria | |
| Powder | |
| Difco |