missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Difco™ Yeast Extract, low-dusting (LD)
GREENER_CHOICE

Catalog No. 210941
Change view
Click to view available options
Quantity:
500 g
10 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
210941 10 kg
DF210933 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 210941 Supplier Gibco™ Supplier No. 210941

Formulation of culture media for cultivation of heterotrophic microorganisms

  • Yeast extract designed to limit the amount of dust generated compared to other spray-dried yeast extracts
  • Yeast Extracts are concentrates of the water-soluble portion of autolyzed Saccharomyces cerevisiae cells
  • Gibco™ yeast extracts are derived from primary grown baker’s yeast
  • These animal origin-free products are suitable for use as multi-functional nutritional supplements in mammalian cell culture, microbial fermentation, and insect cell culture applications
  • Yeast extract provides essential water-soluble vitamins, amino acids, peptides, and carbohydrates to any medium formulation

Specifications

Certifications/Compliance This product does not contains HAPs; This product does not contain any Proposition 65 chemicals; WHMIS Hazard Class - Non-controlled - This product has been classified in accordance with the hazard criteria of the Controlled Products Regulations (CPR)and the SDS contains all the information required by the CPR
Content And Storage Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers.
Cell Type Bacteria
Culture Type Mammalian, Insect, Microbial
Form Powder
Product Type Dehydrated Culture Media
For Use With (Application) Mammalian, Microbial, Vaccine, Insect cell culture applications
Product Line Difco
Quantity 10 kg
Source Yeast, animal origin-free