missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Difco™ Yeast Extract, low-dusting (LD)
GREENER_CHOICE

Catalog No. DF210933
Change view
Click to view available options
Quantity:
500 g
10 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF210933 500 g
210941 10 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. DF210933 Supplier Gibco™ Supplier No. 210933

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Formulation of culture media for cultivation of heterotrophic microorganisms

  • Yeast extract designed to limit the amount of dust generated compared to other spray-dried yeast extracts
  • Yeast Extracts are concentrates of the water-soluble portion of autolyzed Saccharomyces cerevisiae cells
  • Gibco™ yeast extracts are derived from primary grown baker’s yeast
  • These animal origin-free products are suitable for use as multi-functional nutritional supplements in mammalian cell culture, microbial fermentation, and insect cell culture applications
  • Yeast extract provides essential water-soluble vitamins, amino acids, peptides, and carbohydrates to any medium formulation

Specifications

Certifications/Compliance LUO
Content And Storage Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers.
Cell Type Bacteria
Culture Type Mammalian, Insect, Microbial
Form Powder
Product Type Dehydrated Culture Media
Format Bottle
For Use With (Application) Formulation of culture media for the cultivation of heterotrophic microorganisms
Product Line Difco
Quantity 500 g
Source Yeast, animal origin-free
Show More Show Less