missing translation for 'onlineSavingsMsg'
Learn More

Diethylphosphine oxide, Thermo Scientific Chemicals

Numéro de catalogue. AC466700010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
AC466700010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. AC466700010 Fournisseur Thermo Scientific Chemicals Code fournisseur 466700010
Il en reste null

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Identifiants chimiques

CAS 7215-33-0
Molecular Formula C4H11OP
Molecular Weight (g/mol) 106.1

Spécifications

Quantity 1 g
Formula Weight 106.1
Chemical Name or Material Diethylphosphine oxide

RUO – Research Use Only