missing translation for 'onlineSavingsMsg'
Learn More

Diethylenetriamine 98.0+%, TCI America™

Catalog No. D0493500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0493500ML 500 mL
D049325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D0493500ML Supplier TCI America Supplier No. D0493500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 111-40-0
Molecular Formula C4H13N3
Molecular Weight (g/mol) 103.17
MDL Number MFCD00008171
InChI Key RPNUMPOLZDHAAY-UHFFFAOYSA-N
Synonym diethylenetriamine, bis 2-aminoethyl amine, 2,2'-diaminodiethylamine, diethylene triamine, barsamide 115, epicure t, ancamine deta, 1,4,7-triazaheptane, 2,2'-iminodiethylamine, n,n-bis 2-aminoethyl amine
PubChem CID 8111
ChEBI CHEBI:30629
IUPAC Name bis(2-aminoethyl)amine
SMILES NCCNCCN

Specifications

Melting Point -35°C
Color Yellow
Boiling Point 209°C
Quantity 500 mL
UN Number 2079
Formula Weight 103.17
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Diethylenetriamine
EINECSNumber (2)-0159
RTECSNumber IE1225000
TSCA Yes