missing translation for 'onlineSavingsMsg'
Learn More

Diethylene glycol dimethyl ether, 99%, stab. with 100ppm BHT

Catalog No. AAA13397AP
Change view
Click to view available options
Quantity:
500 mL
2500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA13397AP 500 mL
AAA133970F 2500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA13397AP Supplier Thermo Scientific Chemicals Supplier No. A13397AP

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 111-96-6 | C6H14O3 | 134.18 g/mol

Diethylene glycol dimethyl ether is used as a solvent in organic reactions due to its stability towards higher pH and its high boiling point. It is particularly involved in reactions utilizing organometallic reagents such as Grignard reactions and metal hydride reductions. It is also a solvent for hydroboration reactions with diborane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethylene glycol dimethyl ether is used as a solvent in organic reactions due to its stability towards higher pH and its high boiling point. It is particularly involved in reactions utilizing organometallic reagents such as Grignard reactions and metal hydride reductions. It is also a solvent for hydroboration reactions with diborane.

Solubility
Miscible with water, ethanol, acetone, acetic acid, glycerine, pyridine and aldehydes. Slightly miscible with ether.

Notes
Hygroscopic. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 111-96-6
Molecular Formula C6H14O3
MDL Number MFCD00008503
InChI Key SBZXBUIDTXKZTM-UHFFFAOYSA-N
Synonym diglyme, bis 2-methoxyethyl ether, 2-methoxyethyl ether, 1-methoxy-2-2-methoxyethoxy ethane, diethylene glycol dimethyl ether, poly-solv, di 2-methoxyethyl ether, dimethyl carbitol, 2,5,8-trioxanonane
PubChem CID 8150
ChEBI CHEBI:46784
IUPAC Name 1-methoxy-2-(2-methoxyethoxy)ethane
SMILES COCCOCCOC

Specifications

Melting Point -64°C
Density 0.944
Boiling Point 161°C to 162°C
Flash Point 51°C (124°F)
Odor Ether-like
Refractive Index 1.408
Linear Formula (CH3OCH2CH2)2O
Quantity 500 mL
UN Number UN3271
Beilstein 1736101
Sensitivity Hygroscopic
Merck Index 14,3165
Solubility Information Miscible with water,ethanol,acetone,acetic acid,glycerine,pyridine and aldehydes. Slightly miscible with ether.
Molecular Weight (g/mol) 134.175
Formula Weight 134.18
Percent Purity 99%
Chemical Name or Material Diethylene glycol dimethyl ether, stabilized with 100ppm BHT
Show More Show Less
Hazard Category H226-H336-H360FD-H373
Hazard Statement GHS H Statement
H360-H226
May damage fertility or the unborn child.
Flammable liquid and vapour.
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P271-P281-P303+P361+P353-P304+P340-P308+P313-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHERS, N.O.S.
EINECSNumber 203-924-4
RTECSNumber KN3339000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only