missing translation for 'onlineSavingsMsg'
Learn More

Diethylene Glycol Dibutyl Ether 98.0+%, TCI America™

Catalog No. B082825ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B082825ML 25 mL
B0828500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B082825ML Supplier TCI America Supplier No. B082825ML

Chemical Identifiers

CAS 112-73-2
Molecular Formula C12H26O3
Molecular Weight (g/mol) 218.337
MDL Number MFCD00009459
InChI Key KZVBBTZJMSWGTK-UHFFFAOYSA-N
Synonym diethylene glycol dibutyl ether, bis 2-butoxyethyl ether, dibutyl carbitol, butyl diglyme, 1-2-2-butoxyethoxy ethoxy butane, 5,8,11-trioxapentadecane, ether, bis butoxyethyl, 2,2'-dibutoxyethyl ether, ether, bis 2-butoxyethyl
PubChem CID 8210
IUPAC Name 1-[2-(2-butoxyethoxy)ethoxy]butane
SMILES CCCCOCCOCCOCCCC

Specifications

Melting Point -60°C
Color Colorless
Boiling Point 255°C
Quantity 25 mL
Formula Weight 218.34
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethylene Glycol Dibutyl Ether
EINECSNumber (2)-3870&(7)-1321
RTECSNumber KN0350000
TSCA Yes