missing translation for 'onlineSavingsMsg'
Learn More

Diethyldithiocarbamic acid, sodium salt trihydrate, ACS reagent

Catalog No. AC424311000
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC424311000 100 g Glass bottle
AC424315000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC424311000 Supplier Thermo Scientific Chemicals Supplier No. 42431 1000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 20624-25-3 | C5H10NNaS2·3H2O | 225.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 20624-25-3
Molecular Formula C5H10NNaS2·3H2O
Molecular Weight (g/mol) 225.31
MDL Number MFCD00150617
InChI Key WWGXHTXOZKVJDN-UHFFFAOYSA-M
Synonym sodium diethyldithiocarbamate trihydrate, dithiocarb trihydrate, cupral, diethyldithiocarbamic acid sodium salt trihydrate, ditiocarb sodium trihydrate, unii-04574exf2c, diethyldithiocarbamate sodium trihydrate, carbamic acid, diethyldithio-, sodium salt, trihydrate, diethyldithiocarbamate sodium salt trihydrate, diethyldithiokarbaman sodny trihydrat czech
PubChem CID 517546
IUPAC Name sodium;N,N-diethylcarbamodithioate;trihydrate
SMILES CCN(CC)C(=S)[S-].O.O.O.[Na+]

Specifications

Melting Point 95.0°C to 98.5°C
Color Gray to White
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula (C2H5)2NCS2Na·3H2O
Quantity 100 g
Beilstein 04, II, 613
Sensitivity (to copper) Passes Test
Merck Index 15, 3421
Solubility Information (in water) Passes test
Formula Weight 225.31
Grade ACS Reagent
Physical Form Crystals or Powder
Chemical Name or Material Diethyldithiocarbamic acid, sodium salt trihydrate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Very toxic to aquatic life.
Precautionary Statement GHS P Statement
Do not eat,drink or smoke when using this product.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if presen
RTECSNumber EZ6550000

RUO – Research Use Only