missing translation for 'onlineSavingsMsg'
Learn More

Diethylcarbamazine Citrate 98.0+%, TCI America™

Catalog No. D189810G
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D189810G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D189810G Supplier TCI America Supplier No. D189810G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1642-54-2
Molecular Formula C16H29N3O8
Molecular Weight (g/mol) 391.42
MDL Number MFCD00039124
InChI Key PGNKBEARDDELNB-UHFFFAOYSA-N
Synonym diethylcarbamazine citrate, loxuran, caritrol, dicarocide, eosinopin, filazine, franocide, franozan, carbam palatabs, ethodryl citrate
PubChem CID 15432
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylic acid; N,N-diethyl-4-methylpiperazine-1-carboxamide
SMILES OC(=O)CC(O)(CC(O)=O)C(O)=O.CCN(CC)C(=O)N1CCN(C)CC1

Specifications

Melting Point 140°C
Color White
Quantity 10 g
UN Number 2811
Formula Weight 391.42
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Diethylcarbamazine Citrate
RTECSNumber TL1225000
TSCA Yes