missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Diethylcarbamazine

Numéro de catalogue. AC464510050
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
AC464510050 5 g
AC464510250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AC464510050 Fournisseur Thermo Scientific Chemicals Code fournisseur 464510050
Il en reste null

CAS: 90-89-1 | C10H21N3O | 199.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Identifiants chimiques

CAS 90-89-1
Molecular Formula C10H21N3O
Molecular Weight (g/mol) 199.30
InChI Key RCKMWOKWVGPNJF-UHFFFAOYSA-N
IUPAC Name N,N-diethyl-4-methylpiperazine-1-carboxamide
SMILES CCN(CC)C(=O)N1CCN(C)CC1

Spécifications

Melting Point 49.0°C to 51.0°C
Quantity 5 g
Solubility Information Solubility aq. soln.: soluble.
Formula Weight 199.29
Chemical Name or Material Diethylcarbamazine
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Harmful if swallowed.

Precautionary Statement GHS P Statement IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Call a POISON CENTER or doctor/physician if you feel unwell.

Wash face, hands and any exposed skin thoroughly after handling.

EINECSNumber 202-023-3

RUO – Research Use Only