missing translation for 'onlineSavingsMsg'
Learn More

Diethylammonium diethyldithiocarbamate, 98%

Catalog No. AAA1045822
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1045822 100 g
AAA1045806 5 g
AAA1045814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1045822 Supplier Thermo Scientific Chemicals Supplier No. A1045822

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1518-58-7 | C9H22N2S2

Diethylammonium diethyldithiocarbamate is used in the synthesis of Dithiocarbamate. It serve as hosts for the specific binding of various metal cations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethylammonium diethyldithiocarbamate is used in the synthesis of Dithiocarbamate. It serve as hosts for the specific binding of various metal cations.

Solubility
Soluble in Chloroform, Methanol.

Notes
Hygroscopic. Store away from water/moisture and oxidizing agents. Incompatible with strong acids, Strong bases.

Chemical Identifiers

CAS 1518-58-7
Molecular Formula C9H22N2S2
MDL Number MFCD00013146

Specifications

Melting Point 81°C to 84°C
Linear Formula (CH3CH2)2NH2+ (CH3CH2)2NCS2-
Quantity 100 g
Sensitivity Hygroscopic
Solubility Information Soluble in Chloroform,Methanol.
Formula Weight 222.42
Percent Purity 98%
Chemical Name or Material Diethylammonium diethyldithiocarbamate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 216-175-3
RTECSNumber EZ5425000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only