missing translation for 'onlineSavingsMsg'
Learn More

Diethyl sulfate, 98%

Catalog No. AAL14291AP
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL14291AP 500 mL
AAL14291AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAL14291AP Supplier Thermo Scientific Chemicals Supplier No. L14291AP

missing translation for 'validMainframeMsg'

This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

CAS: 64-67-5 | C4H10O4S | 154.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 64-67-5
Molecular Formula C4H10O4S
Molecular Weight (g/mol) 154.18
MDL Number MFCD00009099
InChI Key DENRZWYUOJLTMF-UHFFFAOYSA-N
Synonym sulfuric acid, diethyl ester, diethyl sulphate, diethylsulfate, diaethylsulfat, sulfuric acid diethyl ester, diethyl tetraoxosulfate, diaethylsulfat german, unii-k0fo4vfa7i, des van, diethylester kyseliny sirove
PubChem CID 6163
ChEBI CHEBI:34699
IUPAC Name diethyl sulfate
SMILES CCOS(=O)(=O)OCC

Specifications

Melting Point -25°C
Density 1.177
Boiling Point 208°C (decomposition)
Flash Point 104°C (219°F)
Refractive Index 1.4005
Quantity 500 mL
UN Number UN1594
Beilstein 1209714
Merck Index 14,3130
Formula Weight 154.19
Percent Purity 98%
Chemical Name or Material Diethyl sulfate
Show More Show Less
Hazard Category H302+H332-H311-H314-H335-H340-H350
Hazard Statement GHS H Statement
H340-H350-H314-H302-H312-H332
May cause genetic defects.
May cause cancer.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: DIETHYL SULFATE
EINECSNumber 200-589-6
RTECSNumber WS7875000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only