missing translation for 'onlineSavingsMsg'
Learn More

Diéthyle succinate 99,0+%, TCI America™

Numéro de catalogue. S0103500G
Modifier l'affichage
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
S0103500G 500 g
S010325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. S0103500G Fournisseur TCI America Code fournisseur S0103500G

Identifiants chimiques

CAS 123-25-1
Molecular Formula C8H14O4
Molecular Weight (g/mol) 174.196
MDL Number MFCD00009208
InChI Key DKMROQRQHGEIOW-UHFFFAOYSA-N
Synonym diethyl succinate, ethyl succinate, butanedioic acid, diethyl ester, 1,4-diethyl butanedioate, succinic acid, diethyl ester, succinic acid diethyl ester, diethyl ethanedicarboxylate, unii-elp55c13dr, diethylester kyseliny jantarove, fema no. 2377
PubChem CID 31249
IUPAC Name diéthyle butanedioate
SMILES CCOC(=O)CCC(=O)OCC

Spécifications

Color Incolore
Boiling Point 196°C
Quantity 500 g
Formula Weight 174.20
Percent Purity ≥99.0% (GC)
Physical Form Liquide
Chemical Name or Material Diethyl Succinate
EINECSNumber (2)-0848
RTECSNumber WM7400000
TSCA Yes