missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Phthalate, Pharmaceutical Secondary Standard, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111019456
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019456 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019456 Supplier Merck Emd Millipore Supplier No. PHR14761G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Diethyl Phthalate is a phthalic acid ester, commonly synthesized by the condensation of the carboxy groups of phthalic acid with methanol.. It is used as a solvent for fragrance in cosmetic formulations and personal care products.Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Specifications

CAS 84-66-2
Vapor Pressure 1 mmHg (100°C)
Vapor Density 7.66 (vs. air)
Density 1.12 g/mL (at 25°C)
Linear Formula C6H4-1,2-(CO2C2H5)2
MDL Number MFCD00009111
Melting Point -3°C
Boiling Point 298°C to 299°C
Synonym Diethyl phthalate
Quantity 1 g
Formula Weight 222.24
Grade Certified Reference Material
Show More Show Less