missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl oxalate, 99%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Acidity | 0.01% max. (as oxalic acid) |
| Melting Point | -41.0°C |
| Density | 1.0700g/mL |
| Boiling Point | 185.0°C |
| Flash Point | 76°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 98.5% min. (GC) |
| Packaging | Glass bottle |
| Refractive Index | 1.4085 to 1.4105 |
| Linear Formula | C2H5O2CCO2C2H5 |
| Show More |
Safety and Handling
| missing translation for 'healthHazard1' | GHS Signal Word: Danger |
| missing translation for 'healthHazard2' | GHS H Statement Harmful if swallowed. Causes serious eye irritation. |
| missing translation for 'healthHazard3' | GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Call a POISON CENTER or doctor/physician if you feel unwell. Wash face,hands and any exposed skin thoroughly after handling. Wear eye protection/face protection. |
| EINECSNumber | 202-464-1 |
RUO – Research Use Only