missing translation for 'onlineSavingsMsg'
Learn More

Diethyl maleate, 97%

Catalog No. AAL0387630
Change view
Click to view available options
Quantity:
5 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0387630 250 g
AAL0387606 5 g
AAL038760B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0387630 Supplier Thermo Scientific Chemicals Supplier No. L0387630
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 141-05-9 | C8H12O4 | 172.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 141-05-9
Molecular Formula C8H12O4
Molecular Weight (g/mol) 172.18
MDL Number MFCD00009191
InChI Key IEPRKVQEAMIZSS-WAYWQWQTSA-N
Synonym diethyl maleate, ethyl maleate, maleic acid, diethyl ester, diethylmaleate, maleic acid diethyl ester, 2-butenedioic acid z-, diethyl ester, diethyl z-but-2-enedioate, diethyl 2z-but-2-enedioate, unii-g81wqb56ol, ccris 941
PubChem CID 5271566
ChEBI CHEBI:68508
IUPAC Name diethyl (Z)-but-2-enedioate
SMILES CCOC(=O)C=CC(=O)OCC

Specifications

Melting Point -11°C
Density 1.064
Boiling Point 221°C to 224°C
Flash Point 93°C (199°F)
Odor Pleasant
Refractive Index 1.44
Quantity 250 g
Beilstein 1100825
Merck Index 14,3123
Formula Weight 172.18
Percent Purity 97%
Chemical Name or Material Diethyl maleate
Show More Show Less
Hazard Category H227-H317
Hazard Statement GHS H Statement
H317-H227
May cause an allergic skin reaction.
Combustible liquid.
Precautionary Statement P210-P261-P272-P280-P302+P352-P333+P313-P363-P370+P378q-P501c
EINECSNumber 205-451-9
RTECSNumber ON1225000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only