missing translation for 'onlineSavingsMsg'
Learn More

Diethyl L-(+)-Tartrate 98.0+%, TCI America™

Catalog No. T0003500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0003500G 500 g
T000325G 25 g
T0003100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T0003500G Supplier TCI America Supplier No. T0003500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 87-91-2
Molecular Formula C8H14O6
Molecular Weight (g/mol) 206.19
MDL Number MFCD00009143
InChI Key YSAVZVORKRDODB-UHFFFAOYNA-N
Synonym diethyl l-+-tartrate, 2r,3r-diethyl 2,3-dihydroxysuccinate, l +-diethyl l-tartrate, +-diethyl l-tartrate, diethyl l-tartrate, diethyl-l-tartrate, unii-oq72cpy58z, l-+-tartaric acid diethyl ester, diethyl l-tartarate, tartaric acid, diethyl ester, r,r
PubChem CID 6993580
IUPAC Name 1,4-diethyl 2,3-dihydroxybutanedioate
SMILES CCOC(=O)C(O)C(O)C(=O)OCC

Specifications

Color Yellow
Boiling Point 162°C
Quantity 500 g
Formula Weight 206.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl L-(+)-Tartrate
EINECSNumber (2)-1458
TSCA Yes