missing translation for 'onlineSavingsMsg'
Learn More

Diethyl ether, Spectrophotometric Grade, 99+%, inhibitor free

Catalog No. AA40976M1
Change view
Click to view available options
Quantity:
1 L
4 x 1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA40976M1 4 x 1 L
AA40976K2 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AA40976M1 Supplier Thermo Scientific Chemicals Supplier No. 040976M1

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

Solvent For U.V. Spectrophotometric and General Use | CAS: 60-29-7 | C4H10O | 74.12 g/mol

Solvent for U.V. spectrophotometric and it is also important as solvent in production of cellulose plastics such as cellulose acetate. It is also common solvent for Grignard reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 60-29-7
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
MDL Number MFCD00011646
InChI Key RTZKZFJDLAIYFH-UHFFFAOYSA-N
Synonym diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
PubChem CID 3283
ChEBI CHEBI:35702
IUPAC Name ethoxyethane
SMILES CCOCC

Specifications

Melting Point °C to 116°C
Color Colorless
Appearance Colorless liquid
Density 0.713
Quantity 4 x 1 L
Flash Point -45°C (-49°F)
Ignition Point 170°C (338°F)
Odor Sweet
Physical Form Liquid
Refractive Index 1.3525
Assay Percent Range >99%
Vapor Pressure 586.3 hPa (440mm Hg) at 20°C
UN Number 1155
Boiling Point 34.6°C
Solubility Information Immiscible with water,miscible with alcohol.
Formula Weight 74.12
Grade Spectrophotometry
Chemical Name or Material Diethyl ether
Show More Show Less
Hazard Category H224-H302-H305-H335-H336-H373
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P312-P314-P330-P370+P378q-P501c
DOTInformation DOT Class: 3, Packaging Group: I
EINECSNumber 200-467-2
RTECSNumber KI5775000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only