missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Dibutylmalonate 98.0+%, TCI America™

Numéro de catalogue. D118425ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
D118425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. D118425ML Fournisseur TCI America Code fournisseur D118425ML
Il en reste null

Identifiants chimiques

CAS 596-75-8
Molecular Formula C15H28O4
Molecular Weight (g/mol) 272.385
MDL Number MFCD00026842
InChI Key WHKKUUPZLWUOIW-UHFFFAOYSA-N
Synonym diethyl dibutylmalonate, diethyl 2,2-dibutylmalonate, propanedioic acid, dibutyl-, diethyl ester, diethyl di-n-butylmalonate, diethyldibutylmalonate, malonic acid, dibutyl-, diethyl ester, propanedioic acid, 2,2-dibutyl-, 1,3-diethyl ester, 1,3-diethyl 2,2-dibutylpropanedioate, diethyl=dibutylmalonate, acmc-209mf2
PubChem CID 69000
IUPAC Name diethyl 2,2-dibutylpropanedioate
SMILES CCCCC(CCCC)(C(=O)OCC)C(=O)OCC

Spécifications

Color Colorless
Boiling Point 142°C
Quantity 25 mL
Formula Weight 272.39
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl Dibutylmalonate
EINECSNumber (2)-0879
TSCA Yes